Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIP2 Mouse Monoclonal Antibody [clone: NOT AVAILABLE]

Supplier: Enzo Life Sciences
915-062 915-062-100
101106-542EA 464.15 USD InStock
CA101106-542 101106-542
Anti-PIP2 Mouse Monoclonal Antibody [clone: NOT AVAILABLE]
Antibodies
Metabolism of inositol phospholipids by intracellular signaling mediators is fundamental to signal transduction in eukaryotic cells. PI-4,5-P2 (Phosphatidylinositol 4,5-bisphosphate; PIP2) can be synthesized by phosphorylation of PI-4-P by type I phosphatidylinositol phosphate kinase (PIP5K I), or phosphorylation of PI-5-P by type II PIPK (PIP4K II). PI-4,5-P2 regulation of cellular calcium levels involves its hydrolysis by Phospholipase C (PLC) to produce inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG), which serve as second messengers in the import of calcium via IP3-sensitive ion channels and in the activation of PKC, respectively.

Type: Primary
Antigen: PIP2
Clonality: Monoclonal
Clone: NOT AVAILABLE
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty