Would you like to log in again?
Media Bottles, PETG, WHEATON®
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WPBGC0500S WPBGC1000S WPBGC0250S WPBGC2000SB WPBGC0030S WPBGC2000S WPBGC0060S WPBGC0250SB WPBGC1000SB WPBGC0125S
10112-176CS 85.22 USD InStock
10112-176 10112-174 10112-186 10112-172 10112-184 10112-170 10112-182 10112-180 10112-178 10112-168
Media Bottles, PETG, WHEATON®
Bottles Cell Culture Bottles
These bottles are made of eastman tritan eastar copolyester PETG resin for single use labware and medical devices.
  • Ideal for the storage of media, laboratory reagents and biological buffers
  • Graduation markings are molded in for reference filling
  • Have a standard cap
  • Comprised of resins that are free of animal-derived components
  • Suitable for transport and storage, leak resistant
  • Recommended temperature range +70 to –40 °C
The PETG is an amorphous material that displays performance properties including clarity and toughness as well as excellent gas barrier and chemical inertness. The products can be safely sterilized with gamma irradiation without property loss or color shift. They are provided gamma irradiated sterilized to SAL 10-6 with blue caps attached.
Certifications: Meets ISO 10993 and / or USP Class VI biocompatibility requirements and is Food Contact Status compliant.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty