Would you like to log in again?
BSA ELISA Kit, Cygnus Technologies
Supplier: Cygnus Technologies
F030
101120-078EA 1170.77 USD InStock
101120-078
BSA ELISA Kit, Cygnus Technologies
Assays ELISAs
BSA ELISA kit is intended for quantitating bovine serum albumin (BSA).
This highly sensitive assay for BSA has a detection limit of <250 pg/ml. It can be used to optimize purification processes and in routine quality control of in-process streams, as well as in testing the final product.
Delivery information: Kit contains anti-BSA:HRP conjugate, BSA standards set, A-E, TBS wash concentrate, 20X and Anti-BSA coated microtiter strips.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty