Would you like to log in again?
MICRO SPINCOLUMN G-10, PK25
Supplier: HARVARD APPARATUS MS
Micro SpinColumns^ 25-75 ul of solution with sephadex, c18, c8, c4, CN, NH2, PVOH, PHEA, ion exchange (strong and weak), activated charcoal, cellulose, IMAC or empty, packing in various pore sizes, placed in centrifuge to spin out solvents.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICRO SPINCOLUMN G-10, PK25 101166-940 744504 Pack of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty