Would you like to log in again?
Macro Spin Columns, Harvard Apparatus
Supplier: Harvard Apparatus
743901 743905 744309 743904 743903 744302 743900 743902 744303 743906 744308 743907
101167-030PK 1283.08 USD InStock
101167-030 101167-042 101167-032 101167-088 101167-034 101167-100 101167-102 101167-036 101167-038 101167-028 101167-090 101167-040
Macro Spin Columns, Harvard Apparatus
Filters Centrifugal Filters
These macro spin columns are designed for the purification of small samples (10 to 150 µl) in either single column or high-throughput 96-well format with a standard centrifuge.
  • Easy to use
  • High sample recovery and rapid sample preparation time
These spin columns are pre-filled with gel-filtration packing material. The column material binds and purifies the sample according to size and shape, chemical composition, charge or other physiochemical properties.
Applications include DNA purification, protein purification, peptide purification, small molecule and carbohydrate removal , radiolabel removal, nick translation, affinity separation, salt removal, buffer exchange.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty