Would you like to log in again?
MACRO SPINCLM WEAK ANION, PK25
Supplier: HARVARD APPARATUS MS
Macro SpinColumns^ 75-150 ul of solution with sephadex, c18, c8, c4, CN, NH2, PVOH, PHEA, ion exchange (strong and weak), activated charcoal, cellulose, IMAC or empty, packing in various pore sizes, placed in centrifuge to spin out solvents.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MACRO SPINCLM WEAK ANION, PK25 101167-078 744205 Pack of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty