Would you like to log in again?
FAST SPINDIALYZER 500UL
Supplier: HARVARD APPARATUS MS
DIALYZER^ Drop-in re-usable PTFE dialysis chamber. Two sided membrane chamber, with built in magnet.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FAST SPINDIALYZER 500UL 101167-234 740509 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty