Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Arginine hydrochloride 98.5-101.5%

Supplier: Thermo Fisher Scientific
Synonyms: (S)-(+)-Arginine hydrochlorideL-ArgL-Arginine, hydrochloride(S)-2-Amino-5-guanidinopentanoic acid hydrochlorideL(+)-Arginine hydrochlorideL(+)-Arginine monohydrochloride(S)-(+)-Arginine HClH-L-Arg-OH.HClH-Arg-OH.HCl

J11500-36 J11500-A1
101169-510EA 285.37 USD InStock
101169-510 101173-476
L-Arginine hydrochloride 98.5-101.5%
L-Arginine hydrochloride


Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 216 °C (dec.)
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 14,00780
Order Now
Specification Test Results
Assay 98.5- 101.5%
Impurity Content, Organic Volatile Impurities Meets requirements
Impurity Content, Sulphate (SO₄) ≤0.03%
Loss on Drying ≤0.20%
Optical Rotation +21.4° - 23.6° (c=8 in 6 N HCl)
Identification (FTIR) Conforms
Chloride Content 16.5 - 17.1%
Residue on Ignition ≤0.10%
Thin Layer Chromatography Homogeneous
Heavy Metals (Pb) ≤20 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty