Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: L-Glutamic acid(S)-(+)-1-Aminopropane-1,3-dicarboxylic acidL(+)-α-Glutamic acidH-Glu-OH(S)-(+)-Glutamic acid(S)-(+)-α-Glutamic acidL(+)-1-Aminopropane-1,3-dicarboxylic acid(S)-(+)-2-Aminopentanedioic acidL(+)-2-Aminopentanedioic acid

J16215-A7 J16215-A1 J16215-C4
101172-550EA 873.38 USD InStock
101172-550 101170-914 101169-890
L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure
L(+)-Glutamic acid


Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Melting Pt: 189…192 °C (dec.)
Density: 1.525
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 14,04469
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/27/2299827.jpg
Order Now
Specification Test Results
Assay (Unspecified) - Dry Basis 98.5 - 101.5%
Identification (FTIR) Conforms
Loss on Drying ≤0.1%
Optical Rotation (c=10 in 2 N HCl) 31.2° - 32.2°
Arsenic (As) ≤1.5 ppm
Heavy Metals (Pb) ≤1.5 ppm
Lead (Pb) ≤10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty