Would you like to log in again?
LED Light Fixtures
Supplier: Treston
14-9184903 14-9184902 91849005P 91849009P US-91849006P US-91849010P
10117-584EA 310.57 USD InStock
10117-584 10117-574 10117-576 10117-580 10117-582 10117-578
LED Light Fixtures
Lights LED Lights, Lamps and Bulbs
Height and depth adjustable light and balance rails are fitted to the upright tubes and used for suspending lighting units and power tools.
  • Maximum load: 33 lbs.
  • Can hang tools up to 3 lbs.
Ordering information: Installation is required. Assembly videos available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty