Would you like to log in again?
GC Syringes for Thermo Scientific TriPlus RSH Autosampler, Thermo Scientific
Supplier: Thermo Scientific
365F2461 365J2421 365H2161 365I2331 365D0311 365J2411 365C0231 365I2351 365K2811 365D0331 365D0301 365H2151 365Q2121 365F2451 365D0271 365D0261 365I2341 365I2321 365Q2131 365A0241 365N2721 365J2441 365C0251 365C0221 365H2181 365G2311 365F2441 365D0281 365D0321 365B0251 365C6610 365C0241 365Q2141 365H2171 365H2141 365G2321 365G2331
10117-970EA 384.9 USD InStock
10117-970 10117-950 10117-908 10117-948 10117-926 10117-904 10117-906 10117-922 10117-944 10117-946 10117-968 10117-924 10117-962 10117-984 10057-472 10117-964 10117-920 10117-942 10117-980 10117-960 10117-982 10117-916 10117-938 10117-934 10117-956 10117-978 10117-912 10117-914 10117-958 10117-936 10117-974 10117-952 10117-930 10117-932 10117-954 10117-910 10117-976
GC Syringes for Thermo Scientific TriPlus RSH Autosampler, Thermo Scientific
Syringes Syringes for Chromatography
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability, clarity and accuracy in sample introduction. This gives confidence in analytical results, time-after-time.
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover. In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty