Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% (dry basis)

Supplier: Thermo Fisher Scientific
Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diolTromethamine

J22674-A7 J22674-A1 J22674-36
101170-160EA 145.8 USD InStock
101170-160 101170-162 101174-732
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% (dry basis)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)


Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 219…220 °C/10 mm
Melting Pt: 171…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 14,09772
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg
Order Now
Specification Test Results
Assay (Dry Basis) 99.0 - 101.0%
Assay Absorbance (250 nm, 1 M, 1 cm) ≤0.1
Assay Absorbance (290 nm, 40%, 1 cm) ≤0.2
Loss on Drying ≤1%
pH (5%, H₂O) 10.0 - 11.5
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Insoluble Material ≤0.005%
Heavy Metals (Pb) ≤5 ppm
Iron (Fe) ≤5 ppm
pKa (20 °C) 8.30 ±0.15

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty