Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Tryptophan 98.5-101.5% (dry basis)

Supplier: Thermo Fisher Scientific
Synonyms: L-α-Amino-3-indolepropionic acid(S)-2-Amino-3-(3-indolyl)propionic acidH-Trp-OHL-TryptophanTryptophan

J22765-A1 J22765-22
101170-168EA 129.6 USD InStock
101170-168 101174-006
L(-)-Tryptophan 98.5-101.5% (dry basis)
L(-)-Tryptophan


Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 280…285 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 14,09797
Order Now
Specification Test Results
Assay (Dry Basis) 98.5 to 101.5%
Chloride Content ≤0.05%
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content, Organic Impurities Meets the Requirements
Impurity Content Sulphate (SO₄) ≤0.03%
Loss on Drying ≤0.3%
Optical Rotation –29.4 to –32.8° (c=1 in H₂O)
pH (1%, H₂O) 5.5 to 7.0
Heavy Metals (Pb) ≤15 ppm
Iron (Fe) ≤0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty