Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose LE, Ultrapure for molecular biology, for electrophoresis

Supplier: Thermo Fisher Scientific

J32802-30 J32802-36 J32802-14 J32802-A1 J32802-22
101174-042EA 279.14 USD InStock
101174-042 101170-230 101175-234 101176-084 103728-122
Agarose LE, Ultrapure for molecular biology, for electrophoresis
Agarose


Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg
Order Now
Specification Test Results
Form Granular, free-flowing powder
Assay (unspecified) Gel Background Passes test
Assay (unspecified) Gel Point (1.5%) 36 ± 1.5°C
Assay (unspecified) Gel Strength (1%) ≥ 1200 gm/cm2
Assay (unspecified) Gel Strength (1.5%) ≥ 2500 gm/cm2
Comment: EEO (-Mr) 0.05 - 0.13
Comment: Turbidity (1.5%) ≤ 40 Np
DNase Passes test
Residual water ≤ 7%
Residue on ignition ≤ 0.4%
Impurity content: Sulfate (SO4) ≤ 0.14%
Melting Point: Remelt Point (1.5%) 88 ± 1.5°C
RNase Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty