Would you like to log in again?
LB Agar, Ready-Made Powder, Thermo Scientific
Supplier: Thermo Scientific
J75851-A1
101170-306EA 250.48 USD InStock
101170-306
LB Agar, Ready-Made Powder, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
For the propagation of bacteria. Luria-Bertani Agar, off-white free flowing powder.
Contents:
To use: Dissolve 35gm powder per liter of purified water. Autoclave, cool and make plates.
pH(3.5%, H2O): 7.0 ± 0.2
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty