Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agar Noble agar, Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: Agar Agar

J10907-22 J10907-36 J10907-A1
101172-166EA 522.14 USD InStock
101172-166 101170-362 101171-498
Agar Noble agar, Ultrapure
Agar
Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.


Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,00184
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg
Order Now
Specification Test Results
Assay Absorbance (450 nm, 1.5%) ≤0.150
Assay Gel Point (1.5%) 32 - 37.5 °C
Assay Gel Strength (1.5%) ≥700 gm/cm2
Assay Gel pH (1.5%) 5.0 - 7.0
Ash ≤1.6%
Comment EEO (-Mr): ≤0.450 (pH 8.4)
Resistance (1% suspension) ≥20,000 Ohms
Residual water ≤10%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty