Would you like to log in again?
Clostridium histolyticum Collagenase
Supplier: Thermo Scientific
J13820-06 J13820-03
101174-296EA 392.54 USD InStock
101174-296 101170-394
Clostridium histolyticum Collagenase
Enzymes
Collagenase contains a mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes.
  • Enzyme activity: ≥125 units/mg powder (dry basis)
  • Enzyme activity: ≥0.15 Wunsch units/mg (powder dry basis)
One unit liberates 1 μmol of L-Leucine equivalents from collagen in 5 hr at 37 °C, pH 7.5. One unit is equal to 0.0012 Wunsch units.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty