Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Warning
Synonyms: DL-1,4-DithiothreitolDTTcis-2,3-Dihydroxy-1,4-butanedithiol(R*,R*)-1,4-Dimercapto-2,3-butandiol

J15397-03 J15397-14 J15397-22 J15397-06
101169-852EA 1782 USD InStock
101169-852 101170-432 101170-430 101170-860
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure for molecular biology
Dithiothreitol (DTT, Cleland's reagent)
Used as a protective agent for SH groups.


Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 125…130 °C/12 mm
Melting Pt: 40…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
Merck Index: 14,03376
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/19/2299819.jpg
Order Now
Specification Test Results
Assay (SH) ≥99.5%
Assay Absorbance (260 nm, 0.1 M, 1 cm) ≤0.40
Assay Absorbance (280 nm, 0.1 M, 1 cm) ≤0.08
DNase Passes Test
Impurity Content, OxidiSed DTT ≤0.3%
Protease Activity Passes Test
RNase Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty