Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Synonyms: Trizma(R) baseTRIS baseTHAM2-Amino-2-(hydroxymethyl)-1,3-propanediolTris(hydroxymethyl)methylamine2-Amino-2-(hydroxymethyl)propane-1,3-diolTromethamine

J75825-A7 J75825-A9 J75825-36 J75825-A1
101170-654EA 204.37 USD InStock
101170-654 101174-856 101173-310 101375-578
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), Ultrapure for molecular biology
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)


Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 219…220 °C/10 mm
Melting Pt: 171…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 14,09772
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/00/2297600.jpg
Order Now
Specification Test Results
Assay (unspecified) - Dry Basis 99.8 - 100.1%
Assay (unspecified) - Absorbance (250 nm, 1 M, 1 cm) ≤0.1
Assay (unspecified) - Absorbance (290 nm, 40%, 1 cm) ≤0.2
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Insoluble Material ≤0.005%
Water Content (Karl Fischer Titration) ≤1%
Loss on Drying ≤1%
pH (5%, H₂O) 10.0 - 11.5
pKa (20 °C) 8.30 ±0.15
Arsenic (As) ≤1 ppm
Barium (Ba) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Zinc (Zn) ≤1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty