Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate

Supplier: Thermo Fisher Scientific
Synonyms: Adenosine-5'-triphosphoric acid disodium salt trihydrate(((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate5'-ATP·2Na·3H2O

J10585-06 J10585-14 J10585-09 J10585-36 J10585-22
101170-678EA 1686.05 USD InStock
101170-678 101171-478 101174-164 101174-166 101169-718
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) trihydrate


Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MW: 605.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00150754
CAS Number: 51963-61-2
Order Now
Specification Test Results
Assay (UV/vis) (pH 7 or 0.01 N HCl) 100 ±2%
Reference Constants - epsilonmax (pH 7, NRC) 15.4×10³
Reference Constants - epsilon₂₆₀ (pH 7, DBR) 15.4×10³
Reference Constants - lambdamax (pH 7, NRC) 259 nm
Residual Water ≤15%
lambda-max (UV-Vis) (pH 7.0) 259 ±2 nm
Molar Extinction Coefficient (Molar Absorptivity) epsilonmax (pH 7.0) [15.4 ±0.3]×10³
Spectral Ratio (0.01 N HCl) A₂₅₀/A₂₆₀ 0.85 ±0.05
Spectral Ratio (0.01 N HCl) A₂₈₀/A₂₆₀ 0.22 ±0.03
Spectral Ratio (pH 7.0) A₂₅₀/A₂₆₀ 0.80 ±0.03
Spectral Ratio (pH 7.0) A₂₈₀/A₂₆₀ 0.15 ±0.02

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty