Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextran sulfate sodium salt, M.W. 40000 - 50000

Supplier: Thermo Fisher Scientific
Synonyms: Dextran sulfate sodium from DextranDextran sulphate sodium from Dextran

J14489-09 J14489-18 J14489-22
101175-484EA 599 USD InStock
101175-484 101170-838 101175-006
Dextran sulfate sodium salt, M.W. 40000 - 50000
Dextran sulphate sodium salt


Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty