Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-α-Phenylalanine 99+% (dry basis)

Supplier: Thermo Fisher Scientific
Synonyms: DL-Phenylalanine3-Phenyl-DL-alanineH-DL-Phe-OH

J20100-A1
101171-102EA 919.69 USD InStock
101171-102
DL-α-Phenylalanine 99+% (dry basis)
DL-α-Phenylalanine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: ca 266 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Merck Index: 14,07271
Order Now
Specification Test Results
Assay (unspecified) - Ammonia (NH₃) ≤0.03%
Assay (unspecified) - Dry Basis ≥99.0%
Chloride Content ≤0.02%
Identification (FTIR) Conforms
Residue on Ignition ≤0.3%
Loss on Drying ≤0.3%
Thin Layer Chromatography Homogeneous
Arsenic (As) ≤3 ppm
Heavy Metals (Pb) ≤10 ppm
Iron (Fe) ≤50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty