Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific

Supplier: Thermo Scientific
J75893-AE J75893-AP
101171-428EA 98.94 USD InStock
101171-428 101171-430
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific
Buffers Cell Culture Buffers
Use directly to resuspend and/or dilute purified DNA or RNA.

  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test
  • pH(25°C): 7.5 ± 0.1

1X TE solution consisting of 10mM Tris, pH 7.5, and 1mM EDTA.

Solution is 0.2μm filtered and dispensed into vials and/or durable, square Nalgene bottles.

Storage temperature of 1X solution: Ambient

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty