Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycogen (from oysters), Ultrapure

Supplier: Thermo Fisher Scientific

J16445-22 J16445-06 J16445-14
101171-658EA 448.62 USD InStock
101171-658 101169-902 103728-102
Glycogen (from oysters), Ultrapure
Glycogen


Formula: C₂₄H₄₂O₂₁
MW: 666.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081547
CAS Number: 9005-79-2
EINECS: 232-683-8
Order Now
Specification Test Results
Form Light tan powder
Identification (FTIR) Conforms
Loss on Drying (110°C, 4 hrs.) ≤ 15%
Optical Rotation (c=0.2, H2O, dry basis) +170° to 200°
Solution Test (0.25%, 1M NaCl) Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty