Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Malic acid 99+% (by GC)

Supplier: Thermo Fisher Scientific
Danger
Synonyms: L-(–)-Malic Acid(S)-(-)-Hydroxysuccinic acidL(-)-Hydroxy-1,4-butanedioic acid(S)-(-)-Hydroxy-1,4-butanedioic acid(S)-2-hydroxysuccinic acidL(-)-Hydroxysuccinic acidL(-)-Malic acid

J18700-A1 J18700-36
101171-716EA 740.22 USD InStock
101171-716 101174-514
L(-)-Malic acid 99+% (by GC)
L(-)-Malic acid


Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 100…106 °C
Density: 1.6
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064213
CAS Number: 97-67-6
EINECS: 202-601-5
Merck Index: 14,05707
Order Now
Specification Test Results
Assay (GC) ≥99%
Identification (FTIR) Conforms
Water Content (Karl Fischer Titration) ≤1.0%
Optical Rotation (c=5.5 in pyridine) -26.5 to -30.0°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty