Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfate 99+% ACS, Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: Diammonium sulfateSulphuric acid diammonium saltDiammonium sulphateSulfuric acid diammonium salt

J11254-A7 J11254-A1
101172-192EA 251 USD InStock
101172-192 101172-190
Ammonium sulfate 99+% ACS, Ultrapure
Ammonium sulphate


Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: ca 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,00555
Order Now
Specification Test Results
Assay ≥99.0%
Chloride Content ≤5 ppm
Residue on Ignition ≤0.005%
Impurity Content, Nitrate (NO₃) ≤0.001%
Impurity Content, Phosphate (PO₄) ≤5 ppm
Insoluble Material ≤0.005%
pH (5%, H₂O, 25 °C) 5.0 - 6.0
Heavy Metals (Pb) ≤5 ppm
Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty