Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl pyrocarbonate ≥97%, Ultrapure

Supplier: Thermo Fisher Scientific
Warning
Synonyms: Diethyl dicarbonateDEPC (Diethylpyrocarbonate)DEPC

J14710-AE J14710-AC
101170-852EA 527.69 USD InStock
101170-852 101172-448
Diethyl pyrocarbonate ≥97%, Ultrapure
Diethyl pyrocarbonate
A strong, non-specific inhibitor of ribonuclease. Reacts with the N1 of histidine residues in proteins under mild conditions. It can be used to cross-link and modify proteins by ethoxyformylation of NH₂ and imidazole groups in proteins. It reacts with amines and thus cannot be used with Tris buffer.


Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93…94 °C/18 mm
Melting Pt: 69 °C
Density: 1.101
Flash Pt: 69 °C(156 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,07998
Order Now
Specification Test Results
Assay (unspecified) ≥97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty