Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Leucine 98.5-100.5% (dry basis), ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: L-Leucine(S)-(+)-LeucineLeucineLeuH-Leu-OH

J18280-A7 J18280-22 J18280-A1
101171-704EA 345.18 USD InStock
101171-704 101171-706 101172-666
L(+)-Leucine 98.5-100.5% (dry basis), ultrapure
L(+)-Leucine


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: >300 °C
Density: 1.293
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451
Order Now
Specification Test Results
Assay (Dry Basis) 98.5 - 101.5%
Identification (FTIR) Conforms
Residue on Ignition ≤0.4%
Loss on Drying ≤0.2%
Optical Rotation +14.9 - 17.3° (c=4 in 6 N HCl Dry Basis)
Individual Impurities (Thin Layer Chromatography) ≤0.5%
Total Impurities (Thin Layer Chromatography) ≤2.0%
Trace Metal, Heavy Metals (Pb) ≤15 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty