Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride ≥99.0% ACS, Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: HaliteSea saltSaline

J21618-A1 J21618-A7 J21618-A9 J21618-36
101171-820EA 96.6 USD InStock
101171-820 101171-822 101175-162 101172-982
Sodium chloride ≥99.0% ACS, Ultrapure
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now
Specification Test Results
Assay Chlorate and Nitrate (NO₃) ≤0.003%
Assay ≥99.0%
Residual Halide Bromide (Br) ≤0.01%
Residual Halide Iodide (I) ≤0.002%
Impurity Content Phosphate (PO₄) ≤5 ppm
Impurity Content Sulfate (SO₄) ≤0.004%
Insoluble Material ≤0.005%
pH (5%, H₂O, 25 °C) 5.0 - 9.0
Barium (Ba) Passes Test
Calcium (Ca) ≤0.002%
Heavy Metal (Pb) ≤5 ppm
Iron (Fe) ≤2 ppm
Magnesium (Mg) ≤0.001%
Potassium (K) ≤0.005%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty