Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: SorbitolD(-)-Glucitol(Sorbitol)(2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol

J21710-36 J21710-A1 J21710-A7
101175-736EA 67.54 USD InStock
101175-736 101173-910 101172-996
D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure
D(-)-Sorbitol


Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 14,08725
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/21/2299721.jpg
Order Now
Specification Test Results
Assay (Dry Basis) 91.0 to 100.5%
Assay Reducing Sugars ≤0.3%
Impurity Content Limit of Nickel ≤1 ppm
Impurity Content Total Molds and Yeasts ≤100 cfu/g
Water Content (Karl Fischer Titration) ≤1.5%
pH (10%, H₂O) 3.5 - 7.0
Identification (FTIR) Conforms
Identity (A, B) Passes Tests
Total Aerobic Plate Count ≤1000 cfu/g
Residue on Ignition ≤0.1%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty