Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Succinic acid free acid 99+%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1,4-Butanedioic acid

J21900-A7 J21900-A1 J21900-36
101176-046EA 86.48 USD InStock
101176-046 101173-926 101173-004
Succinic acid free acid 99+%, Ultrapure
Succinic acid


Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C
Melting Pt: 186…188 °C
Density: 1.56
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
Merck Index: 14,08869
https://us-legacy.vwr.com/stibo/web/std.lang.all/01/38/2300138.jpg
Order Now
Specification Test Results
Assay (unspecified) ≥99.0%
Water Content (Karl Fischer Titration) ≤1.0%
Identification (FTIR) Conforms
Residue on Ignition ≤0.025%
Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty