Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tricine ≥98.5% (dry basis), ultrapure

Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acidTricine bufferN-(Tris(hydroxymethyl)methyl)glycine

J22561-36 J22561-22 J22561-A1
101173-060EA 62.2 USD InStock
101173-060 101171-190 101173-062
Tricine ≥98.5% (dry basis), ultrapure
Tricine


Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: ca 183 °C (dec.)
Density: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
Merck Index: 14,09651
Order Now
Specification Test Results
Assay, Absorbance (250 nm, 5%, 1 cm) ≤0.1
Assay (Dry Basis) ≥98.5%
Identification (FTIR) Conforms
Loss on Drying ≤0.5%
pKa (20 °C) 8.15 ±0.1
Iron (Fe) ≤30 ppm
Lead (Pb) ≤10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty