Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea ≥99.5%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

J75826-A1 J75826-A7
101173-314EA 461.08 USD InStock
101173-314 101173-312
Urea ≥99.5%, Ultrapure for molecular biology
Urea


Formula: NH₂CONH₂
MW: 60.06 g/mol
Melting Pt: 132…135 °C
Density: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 14,09867
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg
Order Now
Specification Test Results
Assay Cyanate None Detected
Assay ≥99.5%
Chloride Content ≤0.0005%
Conductivity ≤15 µmhos/cm
DNase Passes Test
Identification (FTIR) Conforms
Residue on Ignition ≤0.01%
Impurity Content Sulfate (SO4) ≤0.001%
Insoluble Material ≤0.01%
Protease Activity Passes Test
RNase Passes Test
Spectral Ratio (260 nm, 6 M, 1 cm) ≤0.055
Spectral Ratio (280 nm, 6 M, 1 cm) ≤0.044
Copper (Cu) ≤0.5 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty