Would you like to log in again?
Casein, Hammarsten, Thermo Scientific
Supplier: Thermo Scientific
J12840-P2 J12840-22 J12840-Q1 J12840-P5
101173-516EA 620.58 USD InStock
101173-516 101173-512 101173-514 101171-542
Casein, Hammarsten, Thermo Scientific
Proteins and Peptides
Hammarsten casein is solvent extracted to remove fats from whole casein by the procedure of Hammarsten.
  • Protein: ≥95%
  • Loss on drying: ≤10,0%
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty