Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99% (dry basis), powder, Ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acidHEPES free acid

J16926-A1 J16926-22 J16926-A7 J16926-30
101170-478EA 193.15 USD InStock
101170-478 101175-052 101173-706 101170-476
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99% (dry basis), powder, Ultrapure
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
White crystalline powder


Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Melting Pt: ca 236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 14,04654
https://us-legacy.vwr.com/stibo/web/std.lang.all/38/07/41813807.jpg
Order Now
Specification Test Results
Assay (Unspecified) ≥99.0% (Dry Basis)
Identification (FTIR) Conforms
pKa (20 °C) 7.55 ±0.15
A (250 nm, 1 M, 1 cm) ≤0.2
Loss on Drying ≤0.5%
Residue on Ignition ≤0.1%
Chloride (Cl) ≤0.005%
Phosphate (PO₄) ≤0.001%
Sulfate (SO₄) ≤0.01%
Iron (Fe) ≤5 ppm
Heavy Metals (Pb) ≤10 ppm
Assay (Unspecified) Absorbance (250 nm, 1 M, 1 cm) ≤0.2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty