Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taurine ≥98.5%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Synonyms: 2-Aminoethanesulphonic acid2-Aminoethanesulfonic acid

J75824-A1
101174-114EA 296.58 USD InStock
101174-114
Taurine ≥98.5%, Ultrapure for molecular biology
Taurine


Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 14,09074
Order Now
Specification Test Results
Assay (unspecified) Absorbance (260 nm, 0.5 M, 1 cm) ≤0.2
Assay (unspecified) Absorbance (280 nm, 0.5 M, 1 cm) ≤0.2
Assay (unspecified) Ammonia (NH₃) ≤0.02%
Assay (unspecified) Use Test Functionally Tested for Electrophoresis
Assay (unspecified) ≥98.5%
Chloride Content ≤0.01%
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulfate (SO₄) ≤0.01%
pH (5%, H₂O) 4.0 to 7.0
Solution test (0.5 M, H₂O) Clear and Colorless
Arsenic (As) ≤2 ppm
Heavy Metals ≤10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty