Would you like to log in again?
LB Broth, Ready-Made Powder, Thermo Scientific
Supplier: Thermo Scientific
J75852-30 J75852-A1
101174-120EA 178.2 USD InStock
101174-120 101173-324
LB Broth, Ready-Made Powder, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
For the propagation of bacteria
Luria-Bertani Broth. Off-white free flowing powder.
pH(2.0%, H2O): 7.0 ± 0.2
Contents:
Casein Peptone: 10 gm/L
Yeast Extract: 5 gm/L
Sodium Chloride: 5 gm/L
To use, dissolve 20gm powder per liter of purified water. Autoclave.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty