Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terrific Broth, Ready-Made Powder, Ultrapure, Thermo Scientific

Supplier: Thermo Scientific
J75856-A1
101174-122EA 215.58 USD InStock
101174-122
Terrific Broth, Ready-Made Powder, Ultrapure, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
Provides greater yields of plasmid DNA and recombinant proteins than LB media by allowing for greater cell density.

Contents:
Casein Peptone: 12 g/L
Yeast Extract: 24 g/L
Potassium Phosphate Dibasic: 12.5 g/L
Potassium Phosphate Monobasic: 2.3 g/L

To use:
Add 50.8 gm powder, 4mL glycerol and 996 mL water to make 1 liter medium. Dissolve and autoclave.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty