Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agar

Supplier: Thermo Fisher Scientific
Synonyms: Agar Agar

J10654-Q1 J10654-P2 J10654-P5
101169-724EA 1973.91 USD InStock
101169-724 101174-168 101172-128
Agar
Agar


Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,00184
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg
Order Now
Specification Test Results
Total Ash ≤6.5%
Loss on Drying ≤20.0%
Identity Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty