Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Raffinose pentahydrate

Supplier: Thermo Fisher Scientific
Synonyms: Raffinose, 5-HydrateGossypose pentahydrateMelitose pentahydrate(3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrateD-(+)-Raffinose pentahydrateRaffinose, PentahydrateRaffinose pentahydrateMelitriose pentahydrate

J21060-A1 J21060-36 J21060-22
101174-648EA 248.77 USD InStock
101174-648 101170-082 101174-646
D-(+)-Raffinose pentahydrate
D(+)-Raffinose pentahydrate


Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Boiling Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
Merck Index: 14,08089
Order Now
Specification Test Results
Identification (FTIR) Conforms
Residue on Ignition ≤0.05%
Loss on Drying ≤15.3%
Optical Rotation (c=4 in H₂O and NH₃) +104.9 ±1.3°
Arsenic (As) ≤0.5 ppm
Heavy Metal (Cu) ≤10 ppm
Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty