Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific

Supplier: Thermo Scientific
J75892-AE J75892-K2 J75892-K8
101170-656EA 159.51 USD InStock
101170-656 101171-426 101174-872
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers
Used in Western blotting protocols.

  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test
  • pH(25°C, 20X): 7.4 ± 0.1

20X TBS (Tris Buffered Saline) solution consisting of 500mM Tris, 60mM KCl and 2.8M NaCl in high purity dH2O, pH 7.4.

Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles. The 5L size is supplied in an upright carton with a convenient dispensing spout.

Dilute to 1X with high purity water for a solution consisting of 25mM Tris, pH 7.4, 3.0mM KCl and 140mM NaCl.

Storage temperature of 20X solution: Ambient

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty