Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kanamycin sulfate, Ultrapure

Supplier: Thermo Fisher Scientific
Danger
Synonyms: Kanamycin MonosulphateKanamycin AKanamycin Monosulfate

J17924-14 J17924-06
101175-072EA 281.63 USD InStock
101175-072 101175-582
Kanamycin sulfate, Ultrapure
Kanamycin sulphate
A selectable marker for cosmid vectors used in transfection of mammalian cells.

It binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,05281
Order Now
Specification Test Results
Identity (A, B, C) Passes Test
Potency ≥750 µg of C₁₈H₃₆N₄O₁₁/mg powder
Residue on Ignition ≤1.0%
Impurity Content Organic Impurities Meets the Requirements
Loss on Drying ≤4.0%
pH (1%, H₂O) 6.5 - 8.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty