Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98.5% (dry basis), ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: 2-Morpholinoethanesulphonic acid hydrate2-(N-Morpholino)ethanesulfonic acid monohydrate2-(N-Morpholino)ethanesulphonic acid monohydrate2-Morpholinoethanesulfonic acid hydrate

J18886-22 J18886-A1
101170-536EA 650.49 USD InStock
101170-536 101175-102
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98.5% (dry basis), ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate


Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: ca 308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 14,05902
Order Now
Specification Test Results
Assay (unspecified) - Absorbance (250 nm, 1 M, 1 cm) ≤0.1
Assay (unspecified) - (dry basis) ≥98.5%
Identification (FTIR) Conforms
Water Content (Karl Fischer Titration) ≤11%
pKa (20 °C) 6.15 ±0.15
Iron (Fe) ≤ 30 ppm
Lead (Pb) ≤ 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty