Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycine 99+%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Synonyms: Aminoacetic acidH-Gly-OH

J16407-A1 J16407-A7 J16407-36
101169-898EA 165.74 USD InStock
101169-898 101175-954 101175-538
Glycine 99+%, Ultrapure for molecular biology
Glycine


Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Melting Pt: ca 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,04491
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg
Order Now
Specification Test Results
Assay (Unspecified) ≥99.0%
Assay (Unspecified) - Hydrolysable Substances Passes Test
Assay (Unspecified) - Readily Carbonisable Substances Passes Test
Chloride Content ≤50 ppm
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulfate (SO₄) ≤50 ppm
lambda-max (UV-Vis) - (1 M, 1 cm ≤0.05) 260 nm
Arsenic (As) ≤1 ppm
Barium (Ba) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Zinc (Zn) ≤1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty