Would you like to log in again?
VortaMix Mini Vortexer, Argos Technologies
Supplier: Argos Technologies
04397-27 04397-28
10118-094EA 311.22 USD InStock
10118-094 10118-092
VortaMix Mini Vortexer, Argos Technologies
Shakers and Mixers Vortex Mixers
The Vortamix Mini Vortexer from Argos Technologies is an unbelievably compact, yet powerful instrument engineered to mix tubes quickly and easily
The standard unit comes with a red rubber gasket cap, but blue, green and purple colored caps are offered separately
Despite its small size and weight, the Vortamix offers an excellent mixing speed of 3,000 rpm.
The vortexing motion is activated by placing a tube on the gasket cap and then pressing down lightly. The innovative design of the gasket cap allows centrifuge and test tubes up to 30 mm in diameter to be mixed with very little effort – including 50mL conical tubes.
Certifications: The Vortamix is CE marked and has and IP rating of DIN EN 60529.
Ordering information: This new instrument comes with a 2 year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty