Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Lipoic acid, high purity

Supplier: VWR
Warning
Synonyms: DL-Thioctic acid (oxidized)(±)​-​1,2-​Dithiolane-​3-​Pentanoic Acid6,8-Dithiooctanoic AcidDL-α-Lipoic AcidDL-6,8-Thioctic AcidLip(S2)

VWRV0494-25G
10118-136EA 371.81 USD InStock
10118-136
(±)-α-Lipoic acid, high purity
(±)-α-Lipoic acid
Boiling point: 160 to 165 °C


Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005474
CAS Number: 1077-28-7
Order Now
Specification Test Results
Identification (IR) PASS
Melting Point 60 - 63 Degrees C
Purity = 99.0 %
Solubility (5%, EtOH) PASS

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty