Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) chloride dihydrate 98-103% ACS

Supplier: VWR
Danger
Synonyms: Stannous chloride dihydrateStannous Chloride Dihydrate

VWRV1B1517-100G VWRV1B1517-500G
10118-156CS 2755.86 USD InStock
10118-156 10118-154
Tin(II) chloride dihydrate 98-103% ACS
Tin (II) chloride dihydrate


Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 12,08939
Order Now
Specification Test Results
Assay 98 - 103%
Calcium = 0.005%
Iron = 0.003%
Lead = 0.01%
Potassium = 0.005%
Sodium = 0.01%
Solubility in hydrochloric acid Pass
Sulfate (P/F) Pass

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty