Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Ornithine hydrochloride ≥99%, Reagent Grade

Supplier: VWR
Synonyms: H-Orn-OH·HClH-Orn-OH Hydrochloride

VWRV0958-500G
10118-204CS 4126.17 USD InStock
10118-204
L(+)-Ornithine hydrochloride ≥99%, Reagent Grade
L(+)-Ornithine hydrochloride


Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6
Order Now
Specification Test Results
Ammonium = 0.1%
Heavy Metals = 10 ppm
Iron = 10 ppm
Loss on Drying = 0.5%
Purity = 99%
Specific Rotation (Dried, c=4, 6N HCl) 23 °- 25 °
Sulfate = 0.02%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty