Would you like to log in again?
Nalgene® Ported Biotainer® Bottle and Carboy Closures, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2560-0490 2560-0489 2560-0489
10118-314CS 176.64 USD InStock
10118-314 10118-324 CA10118-314
Nalgene® Ported Biotainer® Bottle and Carboy Closures, Thermo Scientific
Closures Bottle Closures
Designed to aseptically handle solutions in biopharmaceutical, biotechnology, and laboratory liquid transfer applications, these two- and three-ported closures are designed for use with Nalgene® Biotainer® bottles and carboys. Featuring Animal Derived Component-Free (ADCF) components, including seals that are sonically welded to the cap and leakproof closures, the security of high value contents are guaranteed during fluid transfer.
Closures fit 1L to 20L Biotainer® bottles and carboys in PC and PETG.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty