Would you like to log in again?
FectoPRO® DNA Transfection Kit for Protein Production, Transfection®
Supplier: Polyplus-transfection
FectoPRO®
101000019 101000014 101000019 101000014 101000007 101000007
10118-446EA 15852.36 USD InStock
10118-446 10118-842 CA10118-444 10118-444 CA10118-447 CA10118-842
FectoPRO® DNA Transfection Kit for Protein Production, Transfection®
Transfection Reagents
The FectoPRO® Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various serum-free media, using low DNA amount (\<1 μg/ml of cell culture).
  • Amazing protein and antibody production yields
  • Cost-effective Transient Gene Expression using lower plasmid DNA amount
  • Compatible with various mammalian expression media and cells systems
  • Compliant with biomanufacturing guidelines
  • Reproducible production yields
The FectoPRO®-mediated transfection process is easily scalable from a few milliliters to several liters of cell culture, ensuring robust reproducible protein production.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
FectoPRO® is guaranteed free of animal-origin components, and is chemically defined. Systematic lot management and release testing is performed for each lot produced. Furthermore FectoPRO® transfection kit undergoes advanced quality controls: protein productivity, cell viability, and complete sterility.
Delivery information: Each kit contains equal parts FectoPRO® Reagent and FectoPRO® Booster. FectoPRO® Booster is also available separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty